Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256336.1 | NP_001243265.1 | 3 | 390 | ||
NM_004284.4 | NP_004275.4 | 4 | 497 | |||
NM_024568.2 | NP_078844.2 | 4 | 469 | |||
NR_046070.1 | 4 | 497 | ||||
XM_006711639.3 | XP_006711702.1 | 3 | 365 | |||
XM_017002858.1 | XP_016858347.1 | 5 | 1225 | |||
XM_017002859.1 | XP_016858348.1 | 4 | 352 | |||
XM_017002860.1 | XP_016858349.1 | 4 | 445 | |||
XM_017002861.1 | XP_016858350.1 | 4 | 445 | |||
XM_017002863.1 | XP_016858352.1 | 3 | 260 | |||
XM_017002864.1 | XP_016858353.1 | 4 | 445 | |||
GenBank mRNA | AF054177.1 | - | 4 | 418 | ||
AF537213.1 | - | 4 | 454 | |||
AK001342.1 | - | 4 | 454 | |||
AK027631.1 | - | 4 | 426 | |||
AK223496.1 | - | 2 | 148 | |||
AK298424.1 | - | 3 | 330 | |||
AK307400.1 | - | 4 | 454 | |||
BC001171.1 | - | 4 | 431 | |||
BC005038.1 | - | 4 | 394 | |||
BC043501.1 | - | 4 | 415 | |||
CN255649.1 | - | 1 | 70 | |||
DA007597.1 | - | 4 | 475 | |||
DA155685.1 | - | 4 | 498 | |||
DA745071.1 | - | 4 | 448 | |||
EF560738.1 | - | 4 | 456 | |||
KJ534809.1 | - | 4 | 434 |