Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024571.3 | NP_078847.1 | 1, 2 | 238 | ||
GenBank mRNA | AK026593.1 | - | 1, 2 | 207 | ||
BC001381.1 | - | 1, 2 | 1060 | 3051476 | ||
BC009179.1 | - | 1, 2 | 189 | 3946937 | ||
Z84714.1 | - | 1, 2 | 152 |