Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286562.1 | NP_001273491.1 | 2 | 341 | ||
NM_024573.2 | NP_078849.1 | 3 | 447 | |||
GenBank mRNA | AK022972.1 | - | 3 | 418 | ||
AK298490.1 | - | 2 | 128 | |||
AK310840.1 | - | 2 | 105 | |||
AL110241.1 | - | 3 | 173 | |||
BC011348.1 | - | 3 | 167 | 4340090 |