Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024605.3 | NP_078881.3 | 19 | 1801 | ||
XM_005263215.3 | XP_005263272.1 | 19 | 2037 | |||
XM_017008602.1 | XP_016864091.1 | 20 | 2073 | |||
GenBank mRNA | AB050785.1 | - | 19 | 1801 | ||
AK024549.1 | - | 1 | 101 | |||
AK123034.1 | - | 10 | 833 | |||
AK304458.1 | - | 11 | 984 | |||
AL834360.1 | - | 5 | 442 | |||
BC011920.2 | - | 15 | 1331 | |||
BC047914.1 | - | 5 | 876 | |||
BC106018.1 | - | 2 | 140 | |||
BC109029.2 | - | 19 | 1791 | |||
BC109030.1 | - | 19 | 1790 | |||
BC126899.1 | - | 19 | 1797 | |||
BC128055.1 | - | 19 | 1801 | 40027832 | ||
BX647727.1 | - | 13 | 6931 | |||
CR457328.1 | - | 1 | 100 |