Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018089.2 | NP_001018099.1 | 10, 11 | 2214 | ||
NM_024611.5 | NP_078887.2 | 10, 11 | 2374 | |||
XM_005254661.1 | XP_005254718.1 | 11, 12 | 2373 | |||
XM_005254662.1 | XP_005254719.1 | 11, 12 | 2236 | |||
XM_011522009.1 | XP_011520311.1 | 10, 11 | 2248 | |||
XM_011522010.1 | XP_011520312.1 | 10, 11 | 2117 | |||
XM_011522011.2 | XP_011520313.1 | 9, 10 | 2031 | |||
XM_017022569.1 | XP_016878058.1 | 11, 12 | 2410 | |||
GenBank mRNA | AF502591.1 | - | 10, 11 | 2317 | ||
AK021958.1 | - | 1, 2 | 368 | |||
AK055752.1 | - | 10, 11 | 2317 | |||
AK095389.1 | - | 10, 11 | 2198 | |||
AK127892.1 | - | 3, 4 | 1199 | |||
AL832046.1 | - | 10, 11 | 2341 | |||
BC013684.1 | - | 1, 2 | 369 | |||
BC071685.1 | - | 1, 2 | 263 | |||
BC107427.1 | - | 1, 2 | 347 | |||
BC140894.1 | - | 10, 11 | 2293 | 9021762 | ||
BX647873.1 | - | 10, 11 | 2200 | |||
CR749852.1 | - | 10, 11 | 2167 |