Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000378.4 | NP_000369.3 | 6, 7 | 1385 | ||
NM_001198551.1 | NP_001185480.1 | 7, 8 | 888 | |||
NM_001198552.1 | NP_001185481.1 | 6, 7 | 837 | |||
NM_024424.3 | NP_077742.2 | 7, 8 | 1436 | |||
NM_024426.4 | NP_077744.3 | 7, 8 | 1436 | |||
GenBank mRNA | AB971668.1 | - | 7, 8 | 1595 | ||
AK093168.1 | - | 6, 7 | 833 | |||
AK291736.1 | - | 7, 8 | 1436 | |||
BC032861.2 | - | 7, 8 | 883 | 4827849 | ||
BC046461.1 | - | 7, 8 | 1441 | |||
M30393.1 | - | 6, 7 | 739 | |||
S75264.1 | - | 1, 2 | 208 | |||
X51630.1 | - | 7, 8 | 1422 |