Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184779.1 | NP_001171708.1 | 7, 8 | 1153 | ||
NM_001184780.1 | NP_001171709.1 | 7, 8 | 1280 | |||
NM_024505.3 | NP_078781.3 | 7, 8 | 1237 | |||
NR_033671.2 | 8, 9 | 1468 | ||||
NR_033672.1 | 8, 9 | 1384 | ||||
GenBank mRNA | AF317889.1 | - | 4, 5 | 668 | ||
AF325189.1 | - | 8, 9 | 1359 | |||
AF325190.1 | - | 8, 9 | 1443 | |||
AF353088.1 | - | 7, 8 | 1132 | |||
AF353089.1 | - | 7, 8 | 1216 | |||
AK026011.1 | - | 4, 5 | 722 | |||
AK074058.1 | - | 5, 6 | 906 | |||
AK074071.1 | - | 6, 7 | 4355 | |||
AK314689.1 | - | 7, 8 | 1216 | |||
AY166601.1 | - | 4, 5 | 596 | |||
BC125097.1 | - | 7, 8 | 1146 | 40118286 | ||
BC125098.1 | - | 8, 9 | 1250 | 40118288 | ||
DQ413001.1 | - | 4, 5 | 596 |