Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024520.2 | NP_078796.2 | 3, 4 | 852 | ||
XM_005246852.2 | XP_005246909.1 | 2, 3 | 763 | |||
GenBank mRNA | AK026208.1 | - | 3, 4 | 852 | ||
AL832964.1 | - | 2, 3 | 171 | |||
BC017959.1 | - | 2, 3 | 536 | 4607945 |