Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC52A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253815.1 | NP_001240744.1 | 3 | 993 | ||
NM_001253816.1 | NP_001240745.1 | 3 | 973 | |||
NM_024531.4 | NP_078807.1 | 3 | 1087 | |||
NR_045600.1 | 3 | 1301 | ||||
XM_006716658.2 | XP_006716721.1 | 3 | 1404 | |||
XM_006716659.2 | XP_006716722.1 | 3 | 1219 | |||
XM_006716660.2 | XP_006716723.1 | 3 | 1370 | |||
XM_011517300.2 | XP_011515602.1 | 2 | 764 | |||
XM_017013819.1 | XP_016869308.1 | 3 | 1199 | |||
XM_017013820.1 | XP_016869309.1 | 3 | 934 | |||
XM_017013821.1 | XP_016869310.1 | 3 | 912 | |||
XM_017013822.1 | XP_016869311.1 | 3 | 491 | |||
GenBank mRNA | AB522904.1 | - | 2 | 876 | ||
AK021918.1 | - | 3 | 1257 | |||
AK027888.1 | - | 3 | 966 | |||
AK291581.1 | - | 3 | 993 | |||
AK303822.1 | - | 3 | 892 | |||
AY070774.1 | - | 2 | 809 | |||
BC002917.2 | - | 3 | 1039 | 3954480 |
Set Membership: |
G-protein coupled receptor |