Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024541.2 | NP_078817.2 | 20 | 1529 | ||
XM_006717969.1 | XP_006718032.1 | 20 | 1527 | |||
XM_011540151.2 | XP_011538453.1 | 20 | 1574 | |||
XM_011540152.2 | XP_011538454.1 | 19 | 1461 | |||
XM_011540153.2 | XP_011538455.1 | 19 | 1425 | |||
XM_011540154.2 | XP_011538456.1 | 18 | 1359 | |||
XM_011540156.2 | XP_011538458.1 | 13 | 1493 | |||
XM_011540157.2 | XP_011538459.1 | 19 | 1478 | |||
XM_017016635.1 | XP_016872124.1 | 20 | 1527 | |||
XM_017016636.1 | XP_016872125.1 | 20 | 1527 | |||
GenBank mRNA | AK023176.1 | - | 3 | 158 | ||
AK308479.1 | - | 627 | ||||
BC110511.1 | - | 3 | 158 |