Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRD5A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024592.4 | NP_078868.1 | 4 | 698 | ||
XM_017008601.1 | XP_016864090.1 | 4 | 473 | |||
GenBank mRNA | AK023414.1 | - | 4 | 678 | ||
BC002480.2 | - | 4 | 674 | 3350037 | ||
CR457312.1 | - | 4 | 582 |