Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005498.3 | NP_001005498.2 | 8 | 1208 | ||
NM_024599.5 | NP_078875.4 | 9 | 1370 | |||
XM_005257669.3 | XP_005257726.1 | 9 | 1731 | |||
XM_005257670.1 | XP_005257727.1 | 9 | 1240 | |||
XM_011525249.2 | XP_011523551.1 | 9 | 2687 | |||
XM_011525250.2 | XP_011523552.1 | 10 | 1397 | |||
XM_011525251.2 | XP_011523553.1 | 9 | 1214 | |||
XM_017025079.1 | XP_016880568.1 | 9 | 1644 | |||
GenBank mRNA | AK025994.1 | - | 4 | 394 | ||
AK057882.1 | - | 6 | 1064 | |||
AK074034.1 | - | 7 | 1675 | |||
AK292135.1 | - | 9 | 1363 | |||
AK292166.1 | - | 8 | 1163 | |||
AK303434.1 | - | 7 | 1024 | |||
AL833680.1 | - | 3 | 263 | |||
AY500247.1 | - | 8 | 1059 | |||
BC016034.1 | - | 9 | 1350 | 3910543 | ||
BC035829.1 | - | 4 | 456 |