Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033046.3 | NP_001028218.1 | 7 | 1673 | ||
NM_001100407.2 | NP_001093877.1 | 8 | 1723 | |||
NM_001100408.2 | NP_001093878.1 | 6 | 1631 | |||
NM_001193653.1 | NP_001180582.1 | 8 | 1943 | |||
NM_001193654.1 | NP_001180583.1 | 7 | 1893 | |||
NM_001193655.1 | NP_001180584.1 | 7 | 1837 | |||
NM_001193657.1 | NP_001180586.1 | 8 | 1890 | |||
NR_036514.1 | 6 | 1599 | ||||
NR_036516.1 | 6 | 1550 | ||||
NR_036517.1 | 5 | 1508 | ||||
NR_036518.1 | 4 | 1434 | ||||
XM_006722293.2 | XP_006722356.1 | 6 | 2398 | |||
XM_011523606.2 | XP_011521908.1 | 7 | 2530 | |||
XM_017025072.1 | XP_016880561.1 | 6 | 2578 | |||
XM_017025073.1 | XP_016880562.1 | 5 | 2536 | |||
XM_017025074.1 | XP_016880563.1 | 8 | 1623 | |||
XM_017025075.1 | XP_016880564.1 | 8 | 2392 | |||
XM_017025076.1 | XP_016880565.1 | 9 | 1620 | |||
XM_017025077.1 | XP_016880566.1 | 8 | 1784 | |||
XM_017025078.1 | XP_016880567.1 | 7 | 1581 | |||
GenBank mRNA | AF269290.1 | - | 2 | 1796 | ||
AK054876.1 | - | 7 | 1485 | |||
AK074950.1 | - | 8 | 1820 | |||
AK090484.1 | - | 6 | 3744 | |||
AK092865.1 | - | 4 | 2732 | |||
AK225389.1 | - | 7 | 1579 | |||
BC003595.1 | - | 7 | 1557 | 2822930 | ||
BC004171.2 | - | 7 | 1557 | 2822930 |