Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024540.3 | NP_078816.2 | 6 | 845 | ||
NM_145729.2 | NP_663781.1 | 6 | 886 | |||
XM_011509981.2 | XP_011508283.1 | 6 | 956 | |||
XM_011509982.2 | XP_011508284.1 | 6 | 854 | |||
GenBank mRNA | AK024570.1 | - | 6 | 843 | ||
AK222500.1 | - | 6 | 843 | |||
AK223066.1 | - | 6 | 843 | |||
BC012440.1 | - | 6 | 827 | 3863359 | ||
BC016700.1 | - | 6 | 845 | 4108596 |