Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCPKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040662.1 | NP_001035752.1 | 5 | 964 | ||
NM_024558.2 | NP_078834.2 | 6 | 1069 | |||
NR_049738.1 | 6 | 1058 | ||||
NR_049739.1 | 5 | 1165 | ||||
GenBank mRNA | AK023982.1 | - | 6 | 1058 | ||
AK129564.1 | - | 5 | 1093 | |||
BX247997.1 | - | 5 | 1144 |