Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286562.1 | NP_001273491.1 | 4 | 1099 | ||
NM_024573.2 | NP_078849.1 | 5 | 1205 | |||
GenBank mRNA | AJ420548.1 | - | 1 | 482 | ||
AK022972.1 | - | 5 | 1176 | |||
AK298490.1 | - | 4 | 886 | |||
BC011348.1 | - | 5 | 925 | 4340090 |