Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300746.1 | NP_001287675.1 | 2 | 1266 | ||
NM_001300747.1 | NP_001287676.1 | 3 | 1298 | |||
NM_001300748.1 | NP_001287677.1 | 3 | 1295 | |||
NM_024587.3 | NP_078863.2 | 3 | 1388 | |||
XM_011542138.2 | XP_011540440.1 | 3 | 1517 | |||
GenBank mRNA | AK026006.1 | - | 3 | 1195 | ||
AK310016.1 | - | 3 | 1332 | |||
AL832539.1 | - | 2 | 1100 | |||
BC064520.1 | - | 3 | 1207 | 5582377 |