Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302082.1 | NP_001289011.1 | 5 | 1186 | ||
NM_024603.3 | NP_078879.2 | 5 | 1104 | |||
XM_011542141.2 | XP_011540443.1 | 5 | 3604 | |||
XM_011542142.2 | XP_011540444.1 | 4 | 1052 | |||
XM_017002331.1 | XP_016857820.1 | 6 | 1137 | |||
XM_017002332.1 | XP_016857821.1 | 6 | 1014 | |||
XM_017002334.1 | XP_016857823.1 | 6 | 1233 | |||
GenBank mRNA | AK021650.1 | - | 5 | 1048 | ||
BC007932.2 | - | 5 | 1056 | 4121760 | ||
BC011804.2 | - | 5 | 870 | 4336159 | ||
KU178776.1 | - | 5 | 966 | |||
KU178777.1 | - | 5 | 1048 | |||
KU178778.1 | - | 4 | 832 |