Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320728.1 | NP_001307657.1 | 7 | 1139 | ||
NM_024610.5 | NP_078886.2 | 8 | 1215 | |||
XM_011513130.2 | XP_011511432.1 | 9 | 1133 | |||
XM_011513132.2 | XP_011511434.2 | 8 | 974 | |||
XM_017007179.1 | XP_016862668.1 | 7 | 1039 | |||
XM_017007180.1 | XP_016862669.1 | 4 | 677 | |||
GenBank mRNA | AF400663.1 | - | 8 | 1182 | ||
AK026276.1 | - | 2 | 914 | |||
AK289410.1 | - | 8 | 1215 | |||
BC011897.2 | - | 8 | 1199 | 3949150 | ||
BC017763.1 | - | 2 | 263 |