Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178055.1 | NP_001171526.1 | 27 | 2757 | ||
NM_001178056.1 | NP_001171527.1 | 25 | 2607 | |||
NM_024615.3 | NP_078891.2 | 26 | 2733 | |||
XM_005248596.4 | XP_005248653.1 | 27 | 3113 | |||
XM_011543631.2 | XP_011541933.1 | 27 | 2646 | |||
XM_011543632.2 | XP_011541934.2 | 26 | 2567 | |||
XM_011543634.2 | XP_011541936.1 | 28 | 3242 | |||
XM_011543635.2 | XP_011541937.1 | 28 | 2763 | |||
XM_011543636.2 | XP_011541938.1 | 27 | 2627 | |||
XM_011543640.2 | XP_011541942.1 | 24 | 2517 | |||
XM_011543641.2 | XP_011541943.1 | 25 | 2494 | |||
XM_011543642.2 | XP_011541944.1 | 23 | 2481 | |||
XM_011543643.2 | XP_011541945.1 | 24 | 2922 | |||
XM_017009850.1 | XP_016865339.1 | 26 | 3193 | |||
XM_017009851.1 | XP_016865340.1 | 25 | 2633 | |||
GenBank mRNA | AK024961.1 | - | 16 | 1803 | ||
AK225188.1 | - | 16 | 1803 | |||
AK289369.1 | - | 26 | 2716 | |||
AK298680.1 | - | 25 | 2596 | |||
AL834477.1 | - | 26 | 2692 | |||
BC037386.1 | - | 23 | 2555 | 4830820 | ||
BC075801.1 | - | 25 | 2566 | 5226828 |