Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024629.3 | NP_078905.2 | 9 | 903 | ||
NR_104593.1 | 9 | 903 | ||||
XM_005263218.4 | XP_005263275.2 | 9 | 938 | |||
GenBank mRNA | AF469667.1 | - | 9 | 906 | ||
AF516710.1 | - | 10 | 1320 | |||
AK027121.1 | - | 7 | 713 | |||
AK292297.1 | - | 9 | 849 | |||
BC031520.1 | - | 4 | 251 | |||
BC067802.1 | - | 9 | 843 | |||
BC107744.1 | - | 4 | 348 | |||
BC131556.1 | - | 9 | 903 | 40108113 | ||
BC141854.1 | - | 9 | 901 | 8860370 | ||
CR457376.1 | - | 2 | 107 | |||
DQ907910.1 | - | 9 | 833 | |||
DQ907911.1 | - | 9 | 833 |