Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSANTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308027.1 | NP_001294956.1 | 2 | 728 | ||
NM_001312920.1 | NP_001299849.1 | 1 | 1462 | |||
NM_001312921.1 | NP_001299850.1 | 2 | 585 | |||
NM_024631.3 | NP_078907.2 | 2 | 572 | |||
XM_005271674.3 | XP_005271731.3 | 3 | 1137 | |||
XM_006718905.1 | XP_006718968.1 | 2 | 2097 | |||
XM_006718906.3 | XP_006718969.1 | 2 | 1933 | |||
XM_011542981.2 | XP_011541283.2 | 2 | 1203 | |||
XM_011542982.2 | XP_011541284.2 | 2 | 1203 | |||
XM_017018295.1 | XP_016873784.1 | 2 | 17343 | |||
XM_017018296.1 | XP_016873785.1 | 2 | 2253 | |||
XM_017018297.1 | XP_016873786.1 | 3 | 554 | |||
XM_017018298.1 | XP_016873787.1 | 3 | 13314 | |||
XM_017018299.1 | XP_016873788.1 | 3 | 355 | |||
GenBank mRNA | AK026995.1 | - | 2 | 571 | ||
AK092413.1 | - | 1 | 1136 | |||
AL512691.1 | - | 1 | 1462 | |||
BC064919.1 | - | 2 | 713 | 6093009 | ||
CX781748.1 | - | 2 | 571 | |||
DA379833.1 | - | 2 | 572 |