Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302084.1 | NP_001289013.1 | 3 | 299 | ||
NM_024650.3 | NP_078926.3 | 5 | 591 | |||
GenBank mRNA | AK026184.1 | - | 4 | 392 | ||
AK225435.1 | - | 4 | 392 | |||
AK302025.1 | - | 3 | 251 | |||
BC028240.1 | - | 5 | 594 | 5243907 | ||
BI559574.1 | - | 5 | 507 |