Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024654.4 | NP_078930.3 | 9, 10 | 1751 | ||
XM_005263493.3 | XP_005263550.1 | 7, 8 | 1083 | |||
XM_011542147.2 | XP_011540449.1 | 4, 5 | 677 | |||
XM_017002338.1 | XP_016857827.1 | 7, 8 | 1261 | |||
XM_017002339.1 | XP_016857828.1 | 6, 7 | 977 | |||
GenBank mRNA | AK026976.1 | - | 5, 6 | 761 | ||
AK091284.1 | - | 9, 10 | 1688 | |||
AK289653.1 | - | 9, 10 | 1683 | |||
BC009257.2 | - | 6, 7 | 841 | |||
BC012439.1 | - | 2, 3 | 327 | |||
BC063639.1 | - | 6, 7 | 791 | |||
BC073860.1 | - | 5, 6 | 754 | |||
BC094836.1 | - | 8, 9 | 1126 | |||
BC105095.1 | - | 9, 10 | 1751 | 8144098 | ||
BC110849.1 | - | 9, 10 | 1660 | 6167575 | ||
BC112278.1 | - | 9, 10 | 1751 | 8327746 | ||
BC143646.1 | - | 9, 10 | 1751 |