Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLGALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024656.3 | NP_078932.2 | 10 | 1433 | ||
XM_005260080.4 | XP_005260137.1 | 10 | 1079 | |||
XM_011528297.2 | XP_011526599.1 | 10 | 1071 | |||
XM_011528298.2 | XP_011526600.1 | 10 | 1172 | |||
XM_011528299.2 | XP_011526601.1 | 8 | 946 | |||
GenBank mRNA | AK025982.1 | - | 3 | 211 | ||
AK074941.1 | - | 10 | 1437 | |||
AK075541.1 | - | 10 | 1325 | |||
AK225397.1 | - | 3 | 211 | |||
BC020492.1 | - | 2 | 142 | |||
BC071684.1 | - | 5 | 466 | |||
BC081531.1 | - | 3 | 212 | |||
BC108308.1 | - | 10 | 1388 | 5138787 |