Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243251.1 | NP_001230180.1 | 7 | 655 | ||
NM_024678.5 | NP_078954.4 | 7 | 1136 | |||
XM_011545253.1 | XP_011543555.1 | 7 | 1129 | |||
XM_017018302.1 | XP_016873791.1 | 7 | 1129 | |||
XM_017018303.1 | XP_016873792.1 | 8 | 720 | |||
XM_017018304.1 | XP_016873793.1 | 7 | 648 | |||
GenBank mRNA | AK027094.1 | - | 5 | 444 | ||
AK056980.1 | - | 7 | 1053 | |||
AK225444.1 | - | 5 | 416 | |||
AK315425.1 | - | 7 | 749 | |||
BC007800.2 | - | 7 | 946 | 4128365 |