Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024690.2 | NP_078966.2 | 51 | 40507 | ||
XM_017027486.1 | XP_016882975.1 | 45 | 39587 | |||
XM_017027487.1 | XP_016882976.1 | 44 | 39521 | |||
XM_017027488.1 | XP_016882977.1 | 45 | 39587 | |||
XM_017027489.1 | XP_016882978.1 | 42 | 39433 | |||
XM_017027490.1 | XP_016882979.1 | 40 | 39122 | |||
XM_017027491.1 | XP_016882980.1 | 45 | 39587 | |||
XM_017027492.1 | XP_016882981.1 | 40 | 39119 | |||
XM_017027493.1 | XP_016882982.1 | 40 | 39119 | |||
XM_017027499.1 | XP_016882988.1 | 45 | 30374 | |||
XM_017027500.1 | XP_016882989.1 | 44 | 30293 | |||
XM_017027501.1 | XP_016882990.1 | 24 | 2301 | |||
GenBank mRNA | AF361486.3 | - | 17767 | |||
AF414442.2 | - | 46135 | ||||
AK024365.1 | - | 3 | 250 | |||
AK128681.1 | - | 1 | 886 | |||
AK128856.1 | - | 8 | 927 | |||
AK128857.1 | - | 2203 |