Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318736.1 | NP_001305665.1 | 5 | 437 | ||
NM_001318737.1 | NP_001305666.1 | 4 | 302 | |||
NM_024725.3 | NP_079001.2 | 4 | 302 | |||
XM_005274292.3 | XP_005274349.1 | 2 | 182 | |||
XM_006718911.3 | XP_006718974.1 | 4 | 323 | |||
XM_011542984.2 | XP_011541286.1 | 4 | 966 | |||
XM_011542987.2 | XP_011541289.1 | 5 | 398 | |||
XM_011542988.2 | XP_011541290.1 | 3 | 274 | |||
XM_017018305.1 | XP_016873794.1 | 3 | 2047 | |||
XM_017018306.1 | XP_016873795.1 | 4 | 2131 | |||
XM_017018307.1 | XP_016873796.1 | 5 | 773 | |||
XM_017018308.1 | XP_016873797.1 | 2 | 2007 | |||
XM_017018309.1 | XP_016873798.1 | 2 | 142 | |||
XM_017018310.1 | XP_016873799.1 | 2 | 183 | |||
XM_017018311.1 | XP_016873800.1 | 3 | 2047 | |||
XM_017018312.1 | XP_016873801.1 | 4 | 966 | |||
GenBank mRNA | AF245436.1 | - | 4 | 290 | ||
AK056790.1 | - | 4 | 295 | |||
AK074306.1 | - | 5 | 431 | |||
AK314669.1 | - | 1 | 331 | |||
BC018663.2 | - | 3 | 237 | 2964737 | ||
BC033726.2 | - | 1 | 103 | 5001231 | ||
BP219605.1 | - | 4 | 327 |