Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077186.1 | NP_001070654.1 | 33 | 4507 | ||
NM_001145809.1 | NP_001139281.1 | 34 | 4606 | |||
NM_024729.3 | NP_079005.3 | 32 | 4483 | |||
XM_006723386.3 | XP_006723449.1 | 33 | 4568 | |||
XM_011527320.2 | XP_011525622.1 | 33 | 4585 | |||
XM_011527321.2 | XP_011525623.1 | 32 | 4561 | |||
XM_011527323.2 | XP_011525625.1 | 33 | 4575 | |||
GenBank mRNA | AB111886.1 | - | 23 | 3165 | ||
AB290169.1 | - | 34 | 4576 | |||
AK023943.1 | - | 2 | 147 | |||
AK122704.1 | - | 1 | 1734 | |||
AK303704.1 | - | 3 | 433 | |||
AY165122.1 | - | 33 | 4483 | |||
AY203926.1 | - | 11 | 1548 | |||
BC129803.1 | - | 21 | 2580 | |||
CR936653.1 | - | 3 | 435 |