Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF329 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024620.3 | NP_078896.3 | 4 | 1924 | ||
XM_006723381.2 | XP_006723444.1 | 3 | 1861 | |||
XM_006723382.3 | XP_006723445.1 | 2 | 1870 | |||
XM_006723383.3 | XP_006723446.1 | 2 | 2375 | |||
XM_006723384.3 | XP_006723447.1 | 2 | 2074 | |||
XM_011527310.2 | XP_011525612.1 | 5 | 2308 | |||
XM_011527311.2 | XP_011525613.1 | 4 | 2134 | |||
XM_011527312.2 | XP_011525614.1 | 4 | 1899 | |||
XM_011527313.2 | XP_011525615.1 | 4 | 2211 | |||
XM_011527314.2 | XP_011525616.1 | 4 | 2190 | |||
XM_011527315.1 | XP_011525617.1 | 4 | 2270 | |||
XM_011527316.2 | XP_011525618.1 | 2 | 1750 | |||
XM_017027307.1 | XP_016882796.1 | 4 | 2574 | |||
XM_017027308.1 | XP_016882797.1 | 3 | 2480 | |||
XM_017027309.1 | XP_016882798.1 | 3 | 2235 | |||
XM_017027310.1 | XP_016882799.1 | 3 | 2536 | |||
XM_017027311.1 | XP_016882800.1 | 2 | 2047 | |||
GenBank mRNA | AK022648.1 | - | 1 | 1217 | ||
AK090893.1 | - | 4 | 2270 | |||
AK095349.1 | - | 5 | 2365 | |||
AK096542.1 | - | 2 | 1731 | |||
BC047501.1 | - | 4 | 1921 | 5266873 |