Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005392.3 | NP_005383.3 | 5 | 680 | ||
XM_005252051.2 | XP_005252108.1 | 5 | 674 | |||
XM_006717143.2 | XP_006717206.1 | 5 | 674 | |||
GenBank mRNA | AB014562.2 | - | 4 | 573 | ||
AF043725.1 | - | 5 | 887 | |||
AL834263.1 | - | 5 | 531 | |||
BC152437.1 | - | 4 | 573 |