Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024627.5 | NP_078903.3 | 3 | 1047 | ||
GenBank mRNA | AK024778.1 | - | 1 | 423 | ||
AK225088.1 | - | 1 | 423 | |||
AK291262.1 | - | 3 | 1015 | |||
AL832509.1 | - | 3 | 936 | |||
BC011679.2 | - | 1 | 554 | 4303449 | ||
BX640785.1 | - | 2 | 1136 | |||
BX640969.1 | - | 3 | 1016 | |||
BX640998.1 | - | 3 | 1016 |