Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QTRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256835.1 | NP_001243764.1 | 6, 7 | 894 | ||
NM_001256836.1 | NP_001243765.1 | 3, 4 | 540 | |||
NM_001256837.1 | NP_001243766.1 | 5, 6 | 638 | |||
NM_024638.3 | NP_078914.1 | 7, 8 | 1002 | |||
XM_005247769.3 | XP_005247826.1 | 7, 8 | 783 | |||
XM_005247770.3 | XP_005247827.1 | 5, 6 | 766 | |||
XM_006713744.3 | XP_006713807.1 | 6, 7 | 635 | |||
XM_011513137.2 | XP_011511439.1 | 6, 7 | 865 | |||
XM_017007182.1 | XP_016862671.1 | 6, 7 | 856 | |||
XM_017007183.1 | XP_016862672.1 | 4, 5 | 621 | |||
XM_017007184.1 | XP_016862673.1 | 6, 7 | 856 | |||
GenBank mRNA | AK023022.1 | - | 7, 8 | 892 | ||
AK296938.1 | - | 3, 4 | 493 | |||
AK299313.1 | - | 5, 6 | 616 | |||
AL833709.1 | - | 6, 7 | 862 | |||
BC034559.2 | - | 6, 7 | 876 | 4385906 | ||
BC039265.1 | - | 7, 8 | 973 | 5296335 |