Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLGALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024656.3 | NP_078932.2 | 12 | 1815 | ||
XM_005260080.4 | XP_005260137.1 | 12 | 1461 | |||
XM_011528297.2 | XP_011526599.1 | 12 | 1453 | |||
XM_011528298.2 | XP_011526600.1 | 12 | 1554 | |||
XM_011528299.2 | XP_011526601.1 | 10 | 1328 | |||
GenBank mRNA | AK025982.1 | - | 5 | 593 | ||
AK057140.1 | - | 2 | 1049 | |||
AK074941.1 | - | 12 | 1819 | |||
AK075541.1 | - | 12 | 1707 | |||
AK225397.1 | - | 5 | 593 | |||
BC020492.1 | - | 4 | 524 | |||
BC081531.1 | - | 6 | 692 | |||
BC108308.1 | - | 12 | 1770 | 5138787 |