Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024658.3 | NP_078934.3 | 8, 9 | 895 | ||
NR_051979.1 | 8, 9 | 895 | ||||
GenBank mRNA | AB208897.1 | - | 8, 9 | 785 | ||
AF411122.1 | - | 8, 9 | 766 | |||
AK026991.1 | - | 1, 2 | 70 | |||
AK056527.1 | - | 8, 9 | 816 | |||
AK056702.1 | - | 8, 9 | 742 | |||
AK057044.1 | - | 7, 8 | 853 | |||
AK057277.1 | - | 8, 9 | 799 | |||
AK094107.1 | - | 8, 9 | 744 | |||
AK094897.1 | - | 7, 8 | 902 | |||
AK095306.1 | - | 8, 9 | 743 | |||
AK314305.1 | - | 8, 9 | 774 | |||
BC110804.1 | - | 8, 9 | 797 | 6164334 | ||
BC136759.1 | - | 8, 9 | 846 | 9020749 | ||
BC144399.1 | - | 8, 9 | 846 | |||
BX248267.1 | - | 8, 9 | 777 |