Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256964.1 | NP_001243893.1 | 2, 3 | 460 | ||
NM_001256966.2 | NP_001243895.1 | 2, 3 | 339 | |||
NM_001256967.2 | NP_001243896.1 | 2, 3 | 334 | |||
NM_001256968.2 | NP_001243897.1 | 2, 3 | 231 | |||
NM_001256969.2 | NP_001243898.1 | 2, 3 | 226 | |||
NM_024661.4 | NP_078937.3 | 2, 3 | 352 | |||
XM_011534113.2 | XP_011532415.1 | 3, 4 | 787 | |||
GenBank mRNA | AK022498.1 | - | 2, 3 | 231 | ||
AW957929.1 | - | 2, 3 | 236 | |||
BC011993.1 | - | 1, 2 | 342 | 3857480 | ||
BG336520.1 | - | 2, 3 | 429 | |||
BI755881.1 | - | 2, 3 | 443 | |||
BM810621.1 | - | 2, 3 | 362 | |||
BP307614.1 | - | 2, 3 | 352 | |||
BQ050243.1 | - | 2, 3 | 274 | |||
BQ427785.1 | - | 1, 2 | 236 |