Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005523.5 | NP_005514.1 | 1 | 494 | ||
GenBank mRNA | AK313921.1 | - | 1 | 484 | ||
BC040948.1 | - | 1 | 494 | 5587615 |