Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024686.4 | NP_078962.4 | 11 | 1589 | ||
XM_005271208.2 | XP_005271265.1 | 10 | 1400 | |||
XM_011542159.2 | XP_011540461.1 | 11 | 1779 | |||
XM_017002347.1 | XP_016857836.1 | 12 | 1609 | |||
XM_017002348.1 | XP_016857837.1 | 11 | 1779 | |||
XM_017002349.1 | XP_016857838.1 | 10 | 1364 | |||
GenBank mRNA | AK026686.1 | - | 4 | 355 | ||
AK094174.1 | - | 6 | 1191 | |||
AK126792.1 | - | 11 | 1486 | |||
AK128604.1 | - | 8 | 1568 | |||
AK225574.1 | - | 4 | 355 | |||
AY170843.1 | - | 10 | 1363 | |||
BC028350.2 | - | 4 | 525 | |||
BC032829.1 | - | 10 | 1331 | |||
BC048970.1 | - | 11 | 1531 | 5295193 | ||
BC060878.1 | - | 11 | 1571 | 5270163 | ||
BG718305.1 | - | 4 | 531 |