Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024690.2 | NP_078966.2 | 54 | 40691 | ||
XM_017027486.1 | XP_016882975.1 | 48 | 39771 | |||
XM_017027487.1 | XP_016882976.1 | 47 | 39705 | |||
XM_017027488.1 | XP_016882977.1 | 48 | 39771 | |||
XM_017027489.1 | XP_016882978.1 | 45 | 39617 | |||
XM_017027490.1 | XP_016882979.1 | 43 | 39306 | |||
XM_017027491.1 | XP_016882980.1 | 48 | 39771 | |||
XM_017027492.1 | XP_016882981.1 | 43 | 39303 | |||
XM_017027493.1 | XP_016882982.1 | 43 | 39303 | |||
XM_017027494.1 | XP_016882983.1 | 43 | 39303 | |||
XM_017027497.1 | XP_016882986.1 | 23 | 37428 | |||
XM_017027499.1 | XP_016882988.1 | 48 | 30558 | |||
XM_017027500.1 | XP_016882989.1 | 47 | 30477 | |||
XM_017027501.1 | XP_016882990.1 | 27 | 2485 | |||
GenBank mRNA | AF361486.3 | - | 17951 | |||
AF414442.2 | - | 46319 | ||||
AK024365.1 | - | 6 | 434 | |||
AK128681.1 | - | 4 | 1070 | |||
AK128857.1 | - | 2387 |