Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF385D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024697.2 | NP_078973.1 | 4 | 788 | ||
XM_011534122.1 | XP_011532424.1 | 5 | 769 | |||
XM_011534123.2 | XP_011532425.1 | 5 | 769 | |||
XM_011534124.2 | XP_011532426.1 | 5 | 769 | |||
XM_017007191.1 | XP_016862680.1 | 5 | 769 | |||
XM_017007192.1 | XP_016862681.1 | 5 | 769 | |||
XM_017007193.1 | XP_016862682.1 | 5 | 759 | |||
XM_017007194.1 | XP_016862683.1 | 6 | 824 | |||
XM_017007195.1 | XP_016862684.1 | 4 | 464 | |||
XM_017007196.1 | XP_016862685.1 | 4 | 942 | |||
XM_017007197.1 | XP_016862686.1 | 4 | 1180 | |||
XM_017007198.1 | XP_016862687.1 | 4 | 935 | |||
XM_017007199.1 | XP_016862688.1 | 3 | 290 | |||
XM_017007200.1 | XP_016862689.1 | 3 | 2506 | |||
XM_017007201.1 | XP_016862690.1 | 3 | 311 | |||
XM_017007203.1 | XP_016862692.1 | 5 | 769 | |||
GenBank mRNA | AK026072.1 | - | 4 | 788 | ||
AK310298.1 | - | 5 | 681 | |||
BC007212.1 | - | 4 | 612 | 3347783 |