Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170796.1 | NP_001164267.1 | 3, 4 | 169 | ||
NM_001170797.1 | NP_001164268.1 | 3, 4 | 169 | |||
NM_024699.2 | NP_078975.2 | 3, 4 | 169 | |||
GenBank mRNA | AK024069.1 | - | 3, 4 | 160 | ||
AK225195.1 | - | 3, 4 | 165 | |||
BC022251.2 | - | 3, 4 | 154 | 4695118 | ||
BQ427843.1 | - | 3, 4 | 169 | |||
CR457323.1 | - | 3, 4 | 146 |