Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEMIN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007269.1 | NP_001007270.1 | 2 | 233 | ||
NM_001007270.1 | NP_001007271.1 | 3 | 270 | |||
NM_001319054.1 | NP_001305983.1 | 2 | 212 | |||
NM_001319055.1 | NP_001305984.1 | 2 | 259 | |||
NM_024707.2 | NP_078983.1 | 3 | 356 | |||
XM_005259262.3 | XP_005259319.1 | 3 | 484 | |||
XM_005259263.4 | XP_005259320.1 | 2 | 222 | |||
XM_017027312.1 | XP_016882801.1 | 2 | 459 | |||
GenBank mRNA | AK024018.1 | - | 3 | 340 | ||
AW297699.1 | - | 2 | 260 | |||
AY114106.1 | - | 1 | 119 | |||
BC007793.2 | - | 3 | 328 | 4053402 | ||
BM009097.1 | - | 2 | 214 | |||
BU602037.1 | - | 3 | 359 | |||
CB148876.1 | - | 3 | 272 | |||
CR457321.1 | - | 1 | 119 |
Gene Symbol: LOC105372419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL047654.1 | - | 2 | 490 |