Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024709.4 | NP_078985.3 | 2 | 446 | ||
GenBank mRNA | AK024208.1 | - | 2 | 367 | ||
AK056749.1 | - | 3 | 312 | |||
AK091937.1 | - | 2 | 380 | |||
AK125403.1 | - | 2 | 887 | |||
BC036067.1 | - | 2 | 412 | 5274863 | ||
DA175175.1 | - | 2 | 446 |