Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024713.2 | NP_078989.1 | 2, 3 | 285 | ||
XM_005254668.3 | XP_005254725.1 | 2, 3 | 335 | |||
XM_017022571.1 | XP_016878060.1 | 3, 4 | 369 | |||
XM_017022572.1 | XP_016878061.1 | 3, 4 | 674 | |||
XM_017022573.1 | XP_016878062.1 | 2, 3 | 495 | |||
XM_017022574.1 | XP_016878063.1 | 3, 4 | 348 | |||
XM_017022575.1 | XP_016878064.1 | 2, 3 | 288 | |||
GenBank mRNA | AK026210.1 | - | 1, 2 | 104 | ||
AK293021.1 | - | 2, 3 | 222 | |||
AK297904.1 | - | 2, 3 | 250 | |||
AK311338.1 | - | 2, 3 | 285 | |||
AL136908.1 | - | 2, 3 | 211 | |||
AL833631.1 | - | 2, 3 | 269 | |||
BC026234.1 | - | 2, 3 | 177 | 4693115 | ||
BC111000.1 | - | 2, 3 | 246 | 5751934 |