Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286732.1 | NP_001273661.1 | 4, 5 | 570 | ||
NM_001286733.1 | NP_001273662.1 | 4, 5 | 570 | |||
NM_024719.3 | NP_078995.2 | 4, 5 | 570 | |||
XM_005268333.3 | XP_005268390.1 | 3, 4 | 407 | |||
XM_011537521.2 | XP_011535823.1 | 4, 5 | 566 | |||
XM_011537522.2 | XP_011535824.1 | 4, 5 | 566 | |||
XM_017020755.1 | XP_016876244.1 | 4, 5 | 528 | |||
XM_017020756.1 | XP_016876245.1 | 5, 6 | 708 | |||
GenBank mRNA | AB449887.1 | - | 4, 5 | 473 | ||
AK026127.1 | - | 2, 3 | 213 | |||
AK300897.1 | - | 4, 5 | 520 | |||
BC033071.2 | - | 2, 3 | 294 | |||
BC062461.1 | - | 3, 4 | 418 | |||
BC080640.1 | - | 4, 5 | 527 | |||
BC112123.1 | - | 2, 3 | 247 | 8327591 | ||
BC112125.1 | - | 2, 3 | 259 | 8327593 | ||
BC143682.1 | - | 2, 3 | 247 | |||
BQ689859.1 | - | 4, 5 | 507 | |||
CX788865.1 | - | 3, 4 | 411 |