Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005632.2 | NP_005623.1 | 4 | 1784 | ||
XM_011522620.2 | XP_011520922.1 | 5 | 5083 | |||
XM_011522621.2 | XP_011520923.1 | 5 | 6011 | |||
XM_011522622.1 | XP_011520924.1 | 3 | 4715 | |||
XM_011522623.2 | XP_011520925.1 | 6 | 2786 | |||
XM_011522624.2 | XP_011520926.1 | 5 | 2124 | |||
XM_011522625.2 | XP_011520927.1 | 4 | 2010 | |||
XM_011522626.2 | XP_011520928.1 | 5 | 5575 | |||
XM_011522627.2 | XP_011520929.1 | 5 | 5580 | |||
XM_011522628.2 | XP_011520930.1 | 5 | 2644 | |||
XM_011522629.2 | XP_011520931.1 | 5 | 2507 | |||
XM_011522630.2 | XP_011520932.1 | 4 | 1845 | |||
XM_011522631.2 | XP_011520933.1 | 3 | 1731 | |||
XM_011522632.2 | XP_011520934.1 | 3 | 2162 | |||
XM_017023596.1 | XP_016879085.1 | 6 | 2923 | |||
GenBank mRNA | U85647.1 | - | 4 | 1784 |