Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085354.2 | NP_001078823.1 | 15 | 2640 | ||
NM_024657.4 | NP_078933.3 | 15 | 2640 | |||
XM_005262190.3 | XP_005262247.1 | 14 | 2530 | |||
XM_006724691.2 | XP_006724754.1 | 13 | 2274 | |||
XM_011531027.2 | XP_011529329.1 | 12 | 1928 | |||
XM_017029844.1 | XP_016885333.1 | 14 | 2530 | |||
GenBank mRNA | AK021627.1 | - | 4 | 954 | ||
AK056235.1 | - | 11 | 1852 | |||
AK300305.1 | - | 14 | 2215 | |||
EF125209.1 | - | 15 | 2365 | |||
EF125210.1 | - | 14 | 2424 |