Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077447.2 | NP_001070915.1 | 3 | 874 | ||
NM_001287506.1 | NP_001274435.1 | 3 | 898 | |||
NM_001287507.1 | NP_001274436.1 | 3 | 1115 | |||
NM_001287508.1 | NP_001274437.1 | 3 | 970 | |||
NM_001287509.1 | NP_001274438.1 | 3 | 829 | |||
NM_001287510.1 | NP_001274439.1 | 2 | 1492 | |||
NM_024664.3 | NP_078940.2 | 3 | 1319 | |||
GenBank mRNA | AK021900.1 | - | 3 | 1253 | ||
BC062586.1 | - | 2 | 1442 | |||
BC104938.1 | - | 3 | 1319 | 8143941 | ||
BC106064.1 | - | 3 | 1269 | 3913530 | ||
BC112015.1 | - | 3 | 1319 | 8327483 |