Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAGAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271885.1 | NP_001258814.1 | 10 | 1144 | ||
NM_001271886.1 | NP_001258815.1 | 10 | 1405 | |||
NM_024666.4 | NP_078942.3 | 10 | 1260 | |||
XM_011522020.1 | XP_011520322.1 | 10 | 1510 | |||
GenBank mRNA | AK021568.1 | - | 10 | 1164 | ||
AL136715.1 | - | 10 | 1170 | |||
AL832256.1 | - | 7 | 841 | |||
BC001975.1 | - | 7 | 781 | |||
BC047026.1 | - | 10 | 1183 | 5534878 | ||
BC058886.1 | - | 10 | 1182 | 6503238 | ||
BX648272.1 | - | 1 | 3237 |