Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024690.2 | NP_078966.2 | 79 | 43255 | ||
XM_017027486.1 | XP_016882975.1 | 73 | 42335 | |||
XM_017027487.1 | XP_016882976.1 | 72 | 42269 | |||
XM_017027488.1 | XP_016882977.1 | 71 | 42248 | |||
XM_017027489.1 | XP_016882978.1 | 70 | 42181 | |||
XM_017027490.1 | XP_016882979.1 | 68 | 41870 | |||
XM_017027491.1 | XP_016882980.1 | 68 | 41870 | |||
XM_017027492.1 | XP_016882981.1 | 68 | 41867 | |||
XM_017027493.1 | XP_016882982.1 | 68 | 41867 | |||
XM_017027494.1 | XP_016882983.1 | 68 | 41867 | |||
XM_017027495.1 | XP_016882984.1 | 68 | 41867 | |||
XM_017027496.1 | XP_016882985.1 | 53 | 40466 | |||
XM_017027497.1 | XP_016882986.1 | 48 | 39992 | |||
XM_017027499.1 | XP_016882988.1 | 73 | 33122 | |||
XM_017027500.1 | XP_016882989.1 | 72 | 33041 | |||
XM_017027501.1 | XP_016882990.1 | 52 | 5049 | |||
GenBank mRNA | AF361486.3 | - | 20515 | |||
AF414442.2 | - | 66190 | ||||
AK024365.1 | - | 31 | 2998 | |||
AK128681.1 | - | 23 | 3053 | |||
AK128856.1 | - | 35 | 3624 | |||
AK128857.1 | - | 4885 |